missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-RP2 Matched Antibody Pair, Abnova™
Marke: Abnova H00006102-AP11.1Set
Weitere Details : Netto-Gewicht : 0.00010kg
Beinhaltet: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RP2 (100μg); Detection antibody: mouse monoclonal anti-RP2, IgG2b Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RP2.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Spezifikation
RP2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RP2. | |
DELXp11.3/KIAA0215/TBCCD2 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RP2 (100 μg) 2. Detection antibody: mouse monoclonal anti-RP2, IgG2b Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
RP2 | |
RP2 | |
Affinity Purified | |
RUO | |
6102 | |
Human |