Learn More
Fisherbrand™ General Purpose Digital Heating Circulator
This general purpose digital heated circulator provides great temperature stability and control for demanding applications requiring accurate temperature control.
911.60 € - 1038.80 €
Spezifikation
Kommunikation | Digital |
---|---|
Display | 4-digit LED |
Frequenz | 50/60 Hz |
Produktcode | Marke | Temperaturbereich | Externe Umwälzung | Gewicht (metrisch) | Spannung | Preis | Menge & Verfügbarkeit | ||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Temperaturbereich | Externe Umwälzung | Gewicht (metrisch) | Spannung | Preis | Menge & Verfügbarkeit | ||
17265556
|
Fisherbrand™
|
0°C to 100°C | No | 2.1 kg | 120/230 V |
911.60 €
1 Stück |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
||
17225566
|
Fisherbrand™
|
-20°C to +120°C | Yes | 2.3 kg | 230 V |
1038.80 €
1 Stück |
Verfügbar ab: 24-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||
Beschreibung
- Temperature ranges from ambient 5°C to 100°C (using baths larger than 5 L), to ambient 15°C to 100°C (with 5 L baths)
- Cooling/heating capacities can be reached with a cooling unit
- Simple, intuitive user interface with dial and two function buttons
- Clear, bright 4-digit LED display
- Visual alarm and low liquid detection
- 2-point user calibration
- 3 temperature presets
Applications:
- Clinical, Microbiology and Pathology labs for media tempering, thawing, and incubating samples
- University research for temperature control of spectrophotometers, refractometers and jacketed vessels
- Industrial labs for temperature probe calibration, water analysis, QC testing product, petrochemical testing, material testing, and milk sample testing
Spezifikation
Digital | |
4-digit LED | |
50/60 Hz |