Learn More
TIM-1 Recombinant Rabbit Monoclonal Antibody (152), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen MA530624
Beschreibung
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse TIM-1/KIM-1/HACVR.
CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.Spezifikation
TIM-1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q5QNS5 | |
Havcr1 | |
Recombinant Mouse TIM-1/KIM-1/HACVR Protein (Tyr22-Thr212) | |
100μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA | |
152 | |
Unconjugated | |
Havcr1 | |
AI503787; CD365; HAVCR; HAVCR1; HAVcr-1; Hepatitis A virus cellular receptor 1; hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; Kim1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; t cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain containing 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; TIMD1; TIMD-1 | |
Rabbit | |
Protein A | |
RUO | |
171283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |