missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-5 Monoclonal Antibody (339319), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA524070
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TIM-1, -2, -3, -4, or -6 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
TIM-5 Monoclonal antibody specifically detects TIM-5 in Mouse samples. It is validated for Western Blot.Spezifikation
TIM-5 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Rat | |
Protein A/G | |
RUO | |
Mouse | |
Antibody | |
IgG1 |
Western Blot | |
339319 | |
Unconjugated | |
Gm12169; OTTMUSG00000005497; predicted gene 12169; TIM5; uncharacterized protein LOC210535 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-5 Phe22-Gly161 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |