New Feature
You can now browse the site in English, Italiano, Deutsch or Français.
Sustainability Consumables
Thermo Scientific™ Sustain™ Series PCR Plastics: Reducing Environmental Impact without Sacrificing Performance
July 2025 | 3 min read | By: Thermo Scientific
Thermo Scientific™ Sustain™ Series PCR Plastics: Reducing Environmental Impact without Sacrificing Performance
Introduction
Sustain™ Series PCR Plastics such as those in Figure 1 are designed to deliver accurate and reproducible PCR data while keeping the environment in mind. The biobased polypropylene resin used in their construction is manufactured from a second-generation, bio-circular feedstock (waste and residual oils) under a mass balance approach. Bio-circular feedstocks include materials that are considered waste or processing residue at the beginning of the supply chain, which are not landfilled or energetically used, but instead reused, further used, or recycled in a loop without dropping out of the economy.[1] These feedstocks have a lower environmental footprint compared to virgin fossil fuel-based feedstocks and the mass balance chain-of-custody is certified by the globally recognized International Sustainability and Carbon Certification (ISCC) system.
The Impact of Biobased Materials in Manufacturing
Incorporating biobased material leads to a decrease in the requirement for virgin fossil fuel feedstock, thereby resulting in reduced greenhouse gas emissions. Each kilogram (kg) of biobased polypropylene resin used in these products reduces greenhouse gas emissions by 3.43 kg CO2e.[2] This is equivalent to 1.96x lower CO2e emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based resin. Transitioning to biobased products can thus support customers in meeting their Scope 3 emission reduction targets.[3]
Key Attributes of Sustain™ Series PCR Plastics
As well as the environmental benefits described above, Sustain™ Series PCR Plastics are designed without compromising on product quality or performance. Manufactured in a Class 100,000 cleanroom under ISO 9001 guidelines, the series has several key attributes as follows:
  • Uniform, ultra-thin walls to enable maximum and consistent heat transfer for equally high performance with every sample
  • Visual inspection and electrostatic testing of each well for defects and contaminants. Every lot is run through a PCR cycling test to evaluate sealing
  • Certification as free from DNase, RNase, human DNA, and PCR inhibitors to ensure the accuracy and reproducibility of PCR results
  • Compatibility with Applied Biosystems™ thermal cyclers and a wide range of other popular instruments
A variety of product formats exist to meet specific PCR or qPCR needs, including PCR plates (96-well and 384-well) and strip tubes with attached or separate caps (packaged either together or separately). By choosing Sustain™ Series PCR Plastics, researchers can get accurate, reproducible results while contributing to a greener future and demonstrating a commitment to more responsible sourcing of laboratory plastics through products that really are Greener by Design™.
References 
1 ISCC. Feedstock Category: Bio-Circular. iscc-system.org/markets/feedstocks/bio-circular/
2 Product carbon footprint data provided by the manufacturer of the biobased polypropylene resin. Fossil-based polypropylene has a cradle-to-gate footprint of 1.75 kg CO2e/kg of resin. Biobased polypropylene has a footprint of –1.68 kg CO2e/kg of resin. This includes cradle-to-gate fossil-based emissions (0.96 kg CO2e/kg), biogenic emissions (0.77 kg CO2e/kg), and biogenic removals (–3.41 kg CO2e/kg).
3 Greenhouse Gas Protocol, Corporate Value Chain (Scope 3) Standard. ghgprotocol.org/corporate-value-chain-scope-3-standard