New Feature
You can now browse the site in English, Italiano, Deutsch or Français.
Outcome Certainty Life Sciences
DNA Forensics: Turning Trace Evidence Into Powerful Clues
January 2026 | 3 min read | By: Fisher Scientific Editorial Team
DNA Forensics: Turning Trace Evidence Into Powerful Clues
What Is DNA Forensics?
DNA forensics is the application of DNA analysis to legal and investigative questions. By examining genetic material from blood, saliva, skin cells, hair follicles, or even minute 'touch DNA,' laboratories can generate a DNA profile that is highly specific to an individual.
Since its introduction in the late 1980s, DNA analysis has fundamentally changed criminal investigation work. It contributes to solving cold cases, helps correct miscarriages of justice and supports the identification of victims in disaster situations. Advances in PCR technologies, STR profiling and rapid DNA systems now enable the analysis of significantly smaller or more degraded samples.
From Trace Sample to DNA Profile
1. Collection and Preservation
Collecting samples without contamination is essential. Swabs, gloves, sterile tubes, and proper transport conditions ensure sample integrity. Even minor environmental influences can degrade DNA, which is why documentation and seamless chain-of-custody processes are essential.
2. Extraction and Purification
In the laboratory, the DNA is separated from the sample material and any inhibitors. Modern methods use magnetic particles or spin columns that have been specially developed for forensic applications and enable efficient yields even with degraded or very small sample quantities. 
3. Quantification and Quality Assessment
Prior to STR amplification, the quantity and quality of DNA is determined using real-time PCR. These analyses provide information about:
  • Total amount of human DNA
  • Male-specific DNA
  • Degree of degradation
  • Possible PCR inhibitors
These values support the selection of suitable working parameters such as template quantity or cycle number. 
4. STR Amplification and Profiling
Short tandem repeat (STR) profiling is the global standard in forensic DNA analysis. Specific gene regions are amplified and analysed to generate an individual DNA profile that can be compared with reference samples or matched against approved national databases. 
Quality, Contamination Control, and Standards
Since forensic results have direct legal consequences, laboratories must adhere to strict quality standards.
Key aspects include:
  • Validated methods and reagents for realistic case samples
  • Strict contamination control, e.g. physical separation of pre- and post-PCR areas
  • Documented chain of custody to track every processing step
Compliance with ISO/IEC 17025, the international standard for testing laboratory competence.
These measures ensure the reproducibility, traceability and legal validity of the results.
Emerging Trends in DNA Forensics
Improved Sample Preparation
New extraction chemistries increase the success rate for degraded or difficult trace material.
Advanced Quantification Methods
Modern multiplex PCR kits provide more detailed insights into sample quality and support informed decisions in the STR workflow.
Automation and High Throughput
Automated extraction systems reduce hands-on effort, minimise sources of error and enable laboratories to cope with increasing case numbers.
Innovative Applications
New technologies include:
  • Next-generation sequencing (NGS) for STR and SNP analyses
  • Genealogical forensics
  • Epigenetic age determination
  • Microbiome-based investigation approaches
These methods promise deeper insights from limited or complex trace material.