CAS RN 100017-22-9
CAS RN 100017-22-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
(2R,3S)-1,2-Epoxy-3-hydroxy-4-pentene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R,3S)-1,2-Epoxy-3-hydroxy-4-pentene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2R,3S)-1,2-Epoxy-3-hydroxy-4-pentene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.