CAS RN 10108-87-9
IUPAC Name:
decyltrimethylazanium chloride
Synonyme:
decyltrimethylammonium chloride
Molekulargewicht (g/mol):
235.84
Summenformel:
C13H30ClN
InChi Key:
HXWGXXDEYMNGCT-UHFFFAOYSA-M
SMILES:
[Cl-].CCCCCCCCCC[N+](C)(C)C
1
–
1
von
1
Ergebnisse
1
Decyltrimethylammoniumchlorid, TRC
CAS: 10108-87-9 Summenformel: C13H30N . Cl Molekulargewicht (g/mol): 235.84 Synonym: 1-Decanaminium, N,N,N-trimethyl-, chloride (1:1) (ACI),1-Decanaminium, N,N,N-trimethyl-, chloride (9CI),Ammonium, decyltrimethyl-, chloride (8CI),Decyltrimethylammonium chloride (7CI),Capryltrimethylammonium chloride,n-Decyltrimethylammonium chloride,Trimethylcaprylammonium chloride,Trimethyldecylammonium chloride,Decyl(trimethyl)ammonium chloride SMILES: [Cl-].CCCCCCCCCC[N+](C)(C)C