CAS RN 12039-83-7
CAS RN 12039-83-7
IUPAC Name:
(silanylidenetitanio)silane
Synonyme:
titanium disilicide
Molekulargewicht (g/mol):
108.07
Summenformel:
H4Si2Ti
InChi Key:
WBWPCELMONEWER-UHFFFAOYSA-N
SMILES:
[SiH2]=[Ti]=[SiH2]
1
–
1
von
1
Ergebnisse
1
Titansilizid, 99.5 % (Metallbasis), Thermo Scientific Chemicals
CAS: 12039-83-7 Summenformel: Si2Ti Molekulargewicht (g/mol): 104.037 MDL-Nummer: MFCD00151541 InChI-Schlüssel: DFJQEGUNXWZVAH-UHFFFAOYSA-N Synonym: titanium silicide,titanium disilicide,titanium silicide tisi2,titanium silicide, tisi,titanium silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick,titanium silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick,titanium silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick,titanium silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick PubChem CID: 6336889 IUPAC-Name: Bis($l^{2}-silanyliden)Titan SMILES: [Si]=[Ti]=[Si]