CAS RN 1317-40-4
CAS RN 1317-40-4
IUPAC Name:
sulfanylidenecopper
Synonyme:
copper monosulfide
Molekulargewicht (g/mol):
95.61
Summenformel:
CuS
InChi Key:
BWFPGXWASODCHM-UHFFFAOYSA-N
SMILES:
S=[Cu]
1
–
2
von
2
Ergebnisse
1
Kupfer(II)-sulfid, 99.8 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1317-40-4 Summenformel: CuS Molekulargewicht (g/mol): 95.61 MDL-Nummer: MFCD00016066 InChI-Schlüssel: OMZSGWSJDCOLKM-UHFFFAOYSA-N Synonym: copper ii sulfide,copper monosulfide,cupric sulfide,copper sulfide,copper blue,copper sulphide,oil blue,covellite,copper 2+ sulfide,horace vernet's blue PubChem CID: 14831 SMILES: [S--].[Cu++]
2
Kupfer(II)-sulfid, ultratrocken, 99.98 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1317-40-4 Summenformel: CuS Molekulargewicht (g/mol): 95.61 MDL-Nummer: MFCD00016066 InChI-Schlüssel: OMZSGWSJDCOLKM-UHFFFAOYSA-N Synonym: copper ii sulfide,copper monosulfide,cupric sulfide,copper sulfide,copper blue,copper sulphide,oil blue,covellite,copper 2+ sulfide,horace vernet's blue PubChem CID: 14831 SMILES: [S--].[Cu++]