CAS RN 13760-79-7
CAS RN 13760-79-7
Thulium(III)-fluorid, wasserfrei, 99.9 % (REO), Thermo Scientific Chemicals
CAS: 13760-79-7 Summenformel: F3Tm Molekulargewicht (g/mol): 225.93 MDL-Nummer: MFCD00049611 InChI-Schlüssel: AATUHDXSJTXIHB-UHFFFAOYSA-K IUPAC-Name: thulium(3+) trifluoride SMILES: [F-].[F-].[F-].[Tm+3]
Thulium(III)-fluorid, wasserfrei, REacton™, 99.99 % (REO), Thermo Scientific Chemicals
CAS: 13760-79-7 Summenformel: F3Tm Molekulargewicht (g/mol): 225.93 MDL-Nummer: MFCD00049611 InChI-Schlüssel: AATUHDXSJTXIHB-UHFFFAOYSA-K IUPAC-Name: thulium(3+) trifluoride SMILES: [F-].[F-].[F-].[Tm+3]
Thulium(III)-fluorid, wasserfrei, 99.9 % (REO), Alfa Aesar™
CAS: 13760-79-7 Summenformel: F3Tm Molekulargewicht (g/mol): 225.93 MDL-Nummer: MFCD00049611 InChI-Schlüssel: AATUHDXSJTXIHB-UHFFFAOYSA-K IUPAC-Name: thulium(3+) trifluoride SMILES: [F-].[F-].[F-].[Tm+3]
Thulium(III)-fluorid, ultratrocken, 99.99 % (REO)
CAS: 13760-79-7 Summenformel: F3Tm Molekulargewicht (g/mol): 225.93 MDL-Nummer: MFCD00049611 InChI-Schlüssel: AATUHDXSJTXIHB-UHFFFAOYSA-K IUPAC-Name: thulium(3+) trifluoride SMILES: [F-].[F-].[F-].[Tm+3]