CAS RN 18909-68-7
IUPAC Name:
rubidium(1+) tetrafluoroboranuide
Synonyme:
rubidium(1+) tetrafluoroborate
Molekulargewicht (g/mol):
172.27
Summenformel:
BF4Rb
InChi Key:
MERZTPVDEIGCOQ-UHFFFAOYSA-N
SMILES:
[Rb+].F[B-](F)(F)F
1
–
1
von
1
Ergebnisse
1
Rubidiumtetrafluorborat, 98 %, Thermo Scientific Chemicals
CAS: 18909-68-7 Summenformel: BF4Rb Molekulargewicht (g/mol): 172.27 MDL-Nummer: MFCD00083071 InChI-Schlüssel: MERZTPVDEIGCOQ-UHFFFAOYSA-N Synonym: rubidium tetrafluoroborate,rubidium 1+ tetrafluoroborate,rubidium fluoroborate,rubidium boron fluoride,borate 1-, rubidium,acmc-1bu19,rubidium 1+ ion tetrafluoroborate PubChem CID: 23696894 IUPAC-Name: Rubidium(1+) Tetrafluoroboranuid SMILES: [Rb+].F[B-](F)(F)F