CAS RN 2460022-53-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Tocris Bioscience™ Pomalidomide 4'-alkylC10-amine
Cereblon ligand with alkyl linker and terminal amine for onward chemistry