CAS RN 24621-21-4
IUPAC Name:
niobioamine
Synonyme:
niobium nitride
Molekulargewicht (g/mol):
106.91
Summenformel:
NNb
InChi Key:
CFJRGWXELQQLSA-UHFFFAOYSA-N
SMILES:
N#[Nb]
1
–
1
von
1
Ergebnisse
1
Niob-Nitrid, Thermo Scientific Chemicals
CAS: 24621-21-4 Summenformel: NNb Molekulargewicht (g/mol): 106.913 MDL-Nummer: MFCD00016239 InChI-Schlüssel: CFJRGWXELQQLSA-UHFFFAOYSA-N Synonym: niobium nitride,niobium nitride nbn,columbium nitride,niobiumylidyneamine,niobium nitride 10g PubChem CID: 90560 IUPAC-Name: Azanylidynniob SMILES: N#[Nb]