CAS RN 2467965-70-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Tocris Bioscience™ Pomalidomide 5'-piperidine-acid
Cereblon ligand with piperidine linker and terminal acid for onward chemistry