CAS RN 247069-93-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Tocris Bioscience™ ALDH Detection Reagent - BAAA-DA
Fluorescent ALDH substrate and detection reagent. Used for: identification of cells with high ALDH activity. Application: flow cytometry, fluorescent-activated cell sorting (FACS)