CAS RN 24719-72-0
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Acetylsalicylic Acid Acyl-b-D-glucuronide (>85%), TRC
CAS: 24719-72-0 Summenformel: C15 H16 O10 Molekulargewicht (g/mol): 356.28 Synonym: β-D-Glucopyranuronic acid, 1-[2-(acetyloxy)benzoate],Glucopyranuronic acid, 1-salicylate monoacetate, β-D- (8CI),Salicylic acid acetate, 1-ester with β-D-glucopyranosiduronic acid (8CI) IUPAC-Name: (2S,3S,4S,5R,6S)-6-(2-acetyloxybenzoyl)oxy-3,4,5-trihydroxyoxane-2-carboxylic acid SMILES: CC(=O)Oc1ccccc1C(=O)O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O