CAS RN 25013-15-4
CAS RN 25013-15-4
IUPAC Name:
1-ethenyl-3-methylbenzene
Synonyme:
M-methylstyrene
Molekulargewicht (g/mol):
118.18
Summenformel:
C9H10
InChi Key:
JZHGRUMIRATHIU-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(C=C)=C1