CAS RN 27829-72-7
CAS RN 27829-72-7
IUPAC Name:
ethyl (2E)-hex-2-enoate
Synonyme:
2-hexenoic acid, ethyl ester
Molekulargewicht (g/mol):
142.2
Summenformel:
C8H14O2
InChi Key:
SJRXWMQZUAOMRJ-VOTSOKGWSA-N
SMILES:
CCC\C=C\C(=O)OCC
1
–
1
von
1
Ergebnisse
1
Ethyl-trans-2-Hexenoat, 98 %, Thermo Scientific™
CAS: 27829-72-7 Summenformel: C8H14O2 Molekulargewicht (g/mol): 142.198 MDL-Nummer: MFCD00036541 InChI-Schlüssel: SJRXWMQZUAOMRJ-VOTSOKGWSA-N Synonym: ethyl hex-2-enoate,ethyl 2-hexenoate,ethyl trans-2-hexenoate,2-hexenoic acid, ethyl ester,ethyl e-hex-2-enoate,ethyl e-2-hexenoate,ethyl 2e-hexenoate,2-hexenoic acid, ethyl ester, 2e,unii-ol4euz6c13,2-hexenoic acid, ethyl ester, e PubChem CID: 5364778 IUPAC-Name: Ethyl-(E)-hex-2-enoat SMILES: CCCC=CC(=O)OCC