CAS RN 3685-25-4
CAS RN 3685-25-4
IUPAC Name:
4-aminocyclohexane-1-carboxylic acid
Synonyme:
4-aminocyclohexane-1-carboxylic acid
Molekulargewicht (g/mol):
143.19
Summenformel:
C7H13NO2
InChi Key:
DRNGLYHKYPNTEA-UHFFFAOYSA-N
SMILES:
NC1CCC(CC1)C(O)=O
1
–
1
von
1
Ergebnisse
1
trans-4-Aminocyclohexancarbonsäure, 97 %, Thermo Scientific Chemicals
CAS: 3685-25-4 Summenformel: C7H13NO2 Molekulargewicht (g/mol): 143.186 MDL-Nummer: MFCD00209953 InChI-Schlüssel: DRNGLYHKYPNTEA-UHFFFAOYSA-N Synonym: 4-aminocyclohexanecarboxylic acid,trans-4-aminocyclohexanecarboxylic acid,cis-4-aminocyclohexanecarboxylic acid,cis-4-aminocyclohexane carboxylic acid,cis-4-amino-1-cyclohexanecarboxylic acid,trans-4-amino-cyclohexanecarboxylic acid,cis-4-aminocyclohexanecarboxylicacid,cyclohexanecarboxylic acid, 4-amino-, trans,trans-4-aminocyclohexane carboxylic acid,1s,4s-4-aminocyclohexanecarboxylic acid PubChem CID: 171560 IUPAC-Name: 4-Aminocyclohexan-1-carbonsäure SMILES: C1CC(CCC1C(=O)O)N