CAS RN 5588-84-1
CAS RN 5588-84-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
247.23
Summenformel:
C9H24O4V
InChi Key:
JOUSPCDMLWUHSO-UHFFFAOYSA-N
SMILES:
O=[V].CC(C)O.CC(C)O.CC(C)O
1
–
1
von
1
Ergebnisse
1
Vanadium(V)-triisopropoxid, 96 %, Thermo Scientific Chemicals
CAS: 5588-84-1 Summenformel: C9H24O4V Molekulargewicht (g/mol): 247.23 MDL-Nummer: MFCD00015017 InChI-Schlüssel: JOUSPCDMLWUHSO-UHFFFAOYSA-N Synonym: vanadium v oxytriisopropoxide,oxotris propan-2-olato vanadium,vanadium v triisopropoxide oxide,oxotri isopropoxo vanadium,vtip,vo oipr 3,vanadium triisopropoxy oxide,triisopropoxy oxo vanadium,vanadium v tri-isopropoxy oxide,dsggjxauqhkogq-uhfffaoysa-n PubChem CID: 79702 IUPAC-Name: Oxovanadium; Propan-2-ol SMILES: O=[V].CC(C)O.CC(C)O.CC(C)O