CAS RN 73278-90-7
CAS RN 73278-90-7
IUPAC Name:
methyl 3-[(piperidin-1-yl)methyl]benzoate
Synonyme:
methyl 3-(piperidin-1-ylmethyl)benzoate
Molekulargewicht (g/mol):
233.31
Summenformel:
C14H19NO2
InChi Key:
RJOMIJDQGNNOCO-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(CN2CCCCC2)=CC=C1