CAS RN 74223-64-6
CAS RN 74223-64-6
IUPAC Name:
methyl 2-({[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoyl]amino}sulfonyl)benzoate
Synonyme:
metsulfuron-methyl
Molekulargewicht (g/mol):
381.36
Summenformel:
C14H15N5O6S
InChi Key:
RSMUVYRMZCOLBH-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC1=NC(C)=NC(OC)=N1
1
–
3
von
3
Ergebnisse
2
Metsulfuron-methyl, TRC
CAS: 74223-64-6 Summenformel: C14 H15 N5 O6 S Molekulargewicht (g/mol): 381.36 Synonym: Benzoic acid, 2-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]-, methyl ester,Accurate 60VG,Algrip,Allie SX,Ally,Ally 20DF,Ally XP,Brush-off,Cimarron,DPD 63760H,DPX 6376,DPX-T 6376,Escort,Escort (pesticide),Grench,Gropper,HCHA 92HA,Laren,MSM Turf,Metsulfuron-methyl,N-[(2-Methoxycarbonyl)phenyl]sulfonyl-N'-(6-methoxy-4-methyl-2-triazinyl)urea,Saber DF,Strech 60VP,T 6376,Zinger,Zinger WP IUPAC-Name: methyl 2-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoate SMILES: COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2