Thermo Scientific CO2 Incubators
Delivered with innovative features like HEPA air filtration that surrounds cells with clean room-like air quality and a choice of 100% pure copper or polished stainless-steel interior surfaces. Plus a high-temperature decontamination function that eliminates the need for autoclaving and handling components. The inside story is simple: our CO2 incubators let you culture with complete confidence. Day after day. Year after year.
Enhanced capacity
- Choice of volume capacities ranging from 1.4 cu. ft. to 29 cu. ft.
- Conveniently stackable models for space-constrained labs
- Space to accommodate shakers, stirrers, culture devices or large sample throughput
Quality chamber construction
- Choice of water jacket or direct heat technologies
- Fan-assisted air circulation for uniform environment and rapid recovery
- Choice of reliable in-chamber CO2 measuring technologies
Optimal Growth Environment
- Proven contamination control technologies protect cells
- Variable oxygen control incubators generate hypoxic conditions for healthier growing cells
- Rapid response humidity system prevents cells from drying out
Water Jacket CO2 Incubators
Unique triple wall construction provides unsurpassed temperature stability with dual layers of water and insulation
Product | Capacity | Interior | Shelves | - |
---|---|---|---|---|
6.6 cu. ft. | 18 gauge, cold-rolled steel, powder coated | 4 standard, 17 maximum | Buy Now |
Direct Heat CO2 Incubators
Efficient high performance heaters located on every chamber surface, provide even temperature distribution throughout the entire chamber.
Compact CO2 Incubators
Space savings, small capacity for those seeking a personalized workspace.
Product | Capacity | Interior | Shelves | - |
---|---|---|---|---|
1.4 cu. ft. | Stainless Steel | 3 standard | Buy Now |
Large Capacity CO2 Incubators
Maximum volume for high throughput requirements and easily accommodates shakers, bioreactors and other related equipment.
Product | Capacity | Interior | Shelves | - |
---|---|---|---|---|
29 cu. ft. | Stainless Steel | Adjust on 2 in. (5.1cm) centers | Buy Now |
Variable O2 Control Incubators
Uniquely designed to optimize growth of mammalian cells in an environment that closely mimics in vivo conditions.
Thermo Scientific CO2 Incubators Circulating Fans Flier
Active airflow circulation provides the most uniform growth conditions. Learn more about using proper airflow circulation to achieve optimum results.
Thermo Scientific CO2 Incubators with Variable Oxygen Control Brochure
Learn the five ways to enhance your cell culture in an environment that closely mimics in vivo conditions using a CO2 incubator with variable O2 control.
Thermo Scientific Direct Heat CO2 Incubators Comparison Chart
Compare capacity, heat, filtration, interior and more for five of Thermo Scientific's top direct heat C02 Incubators.
Thermo Scientific Importance of Class 100 Air in a CO2 Incubator White Paper
When an incubator door is opened, laboratory room air enters the chamber. Reducing airborne particles helps minimize the potential damage to your product.
More
Cell Culture Contamination
Understanding the causes and managing the risks.
- Cell Culture Contamination (971.2 KB)
Cell Culture Contamination - Part 2
The role of the CO2 incubator in reducing contamination.
- Cell Culture Contamination - Part 2 (668.4 KB)
Cell Culture Contamination - Part 3
Evaluating contamination prevention solutions for the CO2 incubator.
- Cell Culture Contamination - Part 3 (1.1 MB)