missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
LRRC8A Polyclonal antibody specifically detects LRRC8A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Spezifikation
Spezifikation
| Antigen | LRRC8A |
| Anwendungen | Western Blot, Immunoprecipitation |
| Klassifikation | Polyclonal |
| Konjugat | PerCP |
| Verdünnung | Western Blot, Immunoprecipitation |
| Zusammensetzung | PBS |
| Gen-Alias | AGM5, FLJ10337, FLJ41617, KIAA1437leucine rich repeat containing 8, leucine rich repeat containing 8 family, member A, leucine-rich repeat-containing protein 8A, LRRC8 |
| Wirtsspezies | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 760 to 810 of human Leucine Rich Repeat Containing 8 Family, Member A using the numbering given in entry NP_001120716.1 |
| Reinigungsverfahren | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?