missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dichlor-2-Phenylcyclopropan, 97 %

Catalog Number p-7082015
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2415-80-7
C9H8Cl2
187.063
MFCD00001272
WLWFQGXZIDYWQF-UHFFFAOYSA-N
2,2-dichlorocyclopropyl benzene, 1,1-dichloro-2-phenylcyclopropane, 2,2-dichlorocyclopropyl-benzene, benzene, 2,2-dichlorocyclopropyl, cyclopropane, 1,1-dichloro, 2-phenyl, pubchem18679, acmc-1clka, ksc491m2n, wlwfqgxzidywqf-uhfffaoysa, 2,2-dichloro-cyclopropyl-benzene
95349
(2,2-Dichlorcyclopropyl)benzol
C1C(C1(Cl)Cl)C2=CC=CC=C2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2415-80-7
C9H8Cl2
187.063
MFCD00001272
WLWFQGXZIDYWQF-UHFFFAOYSA-N
2,2-dichlorocyclopropyl benzene, 1,1-dichloro-2-phenylcyclopropane, 2,2-dichlorocyclopropyl-benzene, benzene, 2,2-dichlorocyclopropyl, cyclopropane, 1,1-dichloro, 2-phenyl, pubchem18679, acmc-1clka, ksc491m2n, wlwfqgxzidywqf-uhfffaoysa, 2,2-dichloro-cyclopropyl-benzene
95349
(2,2-Dichlorcyclopropyl)benzol
C1C(C1(Cl)Cl)C2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1,1-Dichloro-2-phenylcyclopropane
Dichte 1.208
Siedepunkt 105°C to 106°C (12 mmHg)
Flammpunkt 104°C (219°F)
Brechungsindex 1.551
Menge 5 g
Beilstein 1860351
Formelmasse 187.07
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'