missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethyl-2-ethenylcyclopropan-1,1-dicarboxylat, TRC

Artikelnummer. 30378215
Change view
Click to view available options
Menge:
100 mg
Unit Size:
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30378215 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378215 Supplier Toronto Research Chemicals Supplier No. TRCD456938100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 1,1-Dimethyl 2-Ethenylcyclopropane-1,1-dicarboxylate
CAS 17447-60-8
Summenformel C9H12O4
Menge 100 mg
Synonym Dimethyl 2-Vinylcyclopropane-1,1-dicarboxylate, Dimethyl 3-Vinylcyclopropane-1,1-dicarboxylate
InChI-Formel InChI=1S/C9H12O4/c1-4-6-5-9(6,7(10)12-2)8(11)13-3/h4,6H,1,5H2,2-3H3
SMILES COC(=O)C1(CC1C=C)C(=O)OC
IUPAC-Name Dimethyl-2-ethenylcyclopropan-1,1-dicarboxylat
Molekulargewicht (g/mol) 184.19
Formelmasse 184.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.