missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthrolin-Eisen(II)-Sulfat, 0.025 M aq. soln., Thermo Scientific Chemicals

Catalog Number p-7049880
Click to view available options
Menge:
50 g
250 g
14634-91-4
C36H24FeN6O4S
692.53
MFCD00036428
CIWXFRVOSDNDJZ-UHFFFAOYSA-L
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
84567
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
14634-91-4
C36H24FeN6O4S
692.53
MFCD00036428
CIWXFRVOSDNDJZ-UHFFFAOYSA-L
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
84567
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

1,10-Phenanthroline iron(II) sulfate is used as an indicator for the iron phenanthroline compound in analytical chemistry. Further, it is used as a redox indicator for making oscillatory Belousov-Zhabotinsky reactions visible.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1,10-Phenanthroline iron(II) sulfate
Farbe Rot
Menge 50 g
IUPAC-Name Eisen(2+);1,10-Phenanthrolin;Sulfat
Formelmasse 692.53
Concentration or Composition (by Analyte or Components) 0.025M wässrige Lösung
Physikalische Form Flüssig

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'