missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-Isopropyliden-Alpha-D-Allofuranose, 98%, Thermo Scientific Chemicals

Catalog Number p-7043232
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2595-05-3
MFCD00135634
CHEBI:34917
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2595-05-3
MFCD00135634
CHEBI:34917

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
Schmelzpunkt 73°C to 76°C
Farbe Weiß
Summenformel C12H17O4PS2
Menge 1 g
Beilstein 1345413
Synonym phenthoate, fenthoate, dimephenthioate, elsan, dimephenthoate, cidial, phendal, dimefenthoat, cidemul, papthion
InChI-Schlüssel XAMUDJHXFNRLCY-UHFFFAOYNA-N
SMILES CCOC(=O)C(C1=CC=CC=C1)SP(=S)(OC)OC
IUPAC-Name Ethyl-2-Dimethoxyphosphinothioylsulfanyl-2-Phenylacetat
Molekulargewicht (g/mol) 320.36
PubChem CID 17435
Formelmasse 260.28
Reinheit (%) 98%
Physikalische Form Pulver
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'