missing translation for 'onlineSavingsMsg'
Learn More

1-[2,5-Bis(trifluormethyl)phenyl]ethanol, 98 %, Thermo Scientific™

Catalog Number p-7031622
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
364-47-6
C10H8F6O
258.163
MFCD10686545
JQSGCKQDLFXCFT-UHFFFAOYSA-N
1-2,5-bis trifluoromethyl phenyl ethanol, 1-2,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-2,5-bis trifluoromethyl
45790936
1-[2,5-bis(trifluormethyl)phenyl]ethanol
CC(C1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
364-47-6
C10H8F6O
258.163
MFCD10686545
JQSGCKQDLFXCFT-UHFFFAOYSA-N
1-2,5-bis trifluoromethyl phenyl ethanol, 1-2,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-2,5-bis trifluoromethyl
45790936
1-[2,5-bis(trifluormethyl)phenyl]ethanol
CC(C1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-[2,5-Bis(trifluoromethyl)phenyl]ethanol
Menge 1 g
Formelmasse 258.16
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'