missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzisoxazol, 97 %, Thermo Scientific Chemicals
Click to view available options
Menge:
1 g
5 g
Beschreibung
1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Löslichkeit
Wasserlöslich. 4.1 mg/ml in Wasser bei 25 °C
Hinweise
Lichtempfindlich. Bei 4 °C lagern. Dunkel lagern. Nicht kompatibel mit Licht und Oxidationsmitteln.
Wasserlöslich. 4.1 mg/ml in Wasser bei 25 °C
Hinweise
Lichtempfindlich. Bei 4 °C lagern. Dunkel lagern. Nicht kompatibel mit Licht und Oxidationsmitteln.
Spezifikation
Chemischer Name oder Material | 1,2-Benzisoxazole |
Dichte | 1.169 |
Siedepunkt | 90°C to 92°C (15 mmHg) |
Flammpunkt | 86°C (186°F) |
Brechungsindex | 1.562 |
Menge | 1 g |
Beilstein | 2154 |
Empfindlichkeit | Light sensitive |
Löslichkeitsinformationen | Soluble in water. 4.1mg/mL in water at 25°C |
Formelmasse | 119.12 |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'