missing translation for 'onlineSavingsMsg'
Learn More

(+)-1,2-Bis-[(2S,5S)-2,5-diethyl-1-phospholanyl]-benzol, ≥ 97 %, Thermo Scientific Chemicals

Catalog Number p-7041042
Click to view available options
Menge:
250 mg
1 g
136779-28-7
C22H36P2
362.478
MFCD00142321
GVVCHDNSTMEUCS-MUGJNUQGSA-N
ethyl-duphos, s,s, unii-fri8026kma, s,s-et-duphos, fri8026kma, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene, +-duphos, 2s,5s-1-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane, s,s-ethyl-duphos
5702640
(2S,5S)-1-[2-[(2S,5S)-2,5-Diethylphospholan-1-yl]Phenyl]-2,5-Diethylphospholan
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC
136779-28-7
C22H36P2
362.478
MFCD00142321
GVVCHDNSTMEUCS-MUGJNUQGSA-N
ethyl-duphos, s,s, unii-fri8026kma, s,s-et-duphos, fri8026kma, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene, +-duphos, 2s,5s-1-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane, s,s-ethyl-duphos
5702640
(2S,5S)-1-[2-[(2S,5S)-2,5-Diethylphospholan-1-yl]Phenyl]-2,5-Diethylphospholan
CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene
Dichte 1.01
Brechungsindex 1.581
Menge 250 mg
Beilstein 4814174
Empfindlichkeit Air Sensitive
Optische Drehung +265° (c=1 in Hexane)
Formelmasse 362.47
Reinheit (%) ≥97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'