missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Di-n-propylaminoethyl)-piperazin, 99 %, Thermo Scientific™

Catalog Number 11430240
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
496808-01-6
C12H27N3
213.37
MFCD00191215
NQRQMDMOABSDEK-UHFFFAOYSA-N
1-2-dipropylamino ethyl piperazine, 1-2-di-n-propylaminoethyl piperazine, 1-2-dipropylaminoethyl piperazine, 1-2-dipropylaminoethyl-piperazine, 1-piperazineethanamine,n,n-dipropyl, 1-2-dipropylamino-ethyl-piperazine, 2-piperazin-1-yl ethyl dipropylamine, n-2-piperazin-1-yl ethyl-n-propylpropan-1-amine, 2-piperazinylethyl dipropylamine
2736329
N-(2-Piperazin-1-ylethyl)-N-Propylpropan-1-Amin
CCCN(CCC)CCN1CCNCC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
496808-01-6
C12H27N3
213.37
MFCD00191215
NQRQMDMOABSDEK-UHFFFAOYSA-N
1-2-dipropylamino ethyl piperazine, 1-2-di-n-propylaminoethyl piperazine, 1-2-dipropylaminoethyl piperazine, 1-2-dipropylaminoethyl-piperazine, 1-piperazineethanamine,n,n-dipropyl, 1-2-dipropylamino-ethyl-piperazine, 2-piperazin-1-yl ethyl dipropylamine, n-2-piperazin-1-yl ethyl-n-propylpropan-1-amine, 2-piperazinylethyl dipropylamine
2736329
N-(2-Piperazin-1-ylethyl)-N-Propylpropan-1-Amin
CCCN(CCC)CCN1CCNCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-(2-Di-n-propylaminoethyl)piperazine
Menge 1 g
Formelmasse 213.37
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'