Learn More
1,2-Diphenylbutan-1-one, Mikromol™
Product Code. 30050268
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050268 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30050268 Supplier Mikromol Supplier No. MM0053.080025
 Restricted Item
Verfügbar ab: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,2-Diphenylbutan-1-one
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2-Diphenylbutan-1-one
CAS 16282-16-9
Analyte or Component Names 1,2-Diphenylbutan-1-one
Shipping Condition Room Temperature
Molecular Formula C16 H16 O
Certifications/Compliance ISO 17025
Synonym 1,2-Diphenylbutan-1-one
InChI Formula InChI=1S/C16H16O/c1-2-15(13-9-5-3-6-10-13)16(17)14-11-7-4-8-12-14/h3-12,15H,2H2,1H3
SMILES CCC(C(=O)c1ccccc1)c2ccccc2
IUPAC Name 1,2-diphenylbutan-1-one
Molecular Weight (g/mol) 224.3
Quantity 25 mg
Formula Weight 224.1201 g/mol