Learn More
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, TRC
Product Code. 30374592
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374592 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30374592 Supplier Toronto Research Chemicals Supplier No. TRCD4361955G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
CAS 69304-37-6
Molecular Formula C12H28Cl2OSi2
Quantity 5 g
Synonym 1,1,3,3-Tetraisopropyl-1,3-dichlorodisiloxane, 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-Dichlorotetraisopropyldisiloxane, TIPDS-Cl2, Tetraisopropyldisiloxane-1,3-diyl Dichloride
InChI Formula InChI=1S/C12H28Cl2OSi2/c1-9(2)16(13,10(3)4)15-17(14,11(5)6)12(7)8/h9-12H,1-8H3
SMILES CC(C)[Si](Cl)(O[Si](Cl)(C(C)C)C(C)C)C(C)C
IUPAC Name chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
Molecular Weight (g/mol) 315.43
Formula Weight 314.1056