missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydrothieno[3,4-b]Chinoxalin, 97 %, Thermo Scientific™

Catalog Number 15465088
Click to view available options
Menge:
2 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3138-80-5
C10H8N2S
188.248
MFCD22683025
OSKBIJDQSJQRQK-UHFFFAOYSA-N
1,3-dihydrothieno 3,4-b quinoxaline, thieno 3,4-b quinoxaline, 1,3-dihydro, 1h,3h-thieno 3,4-b quinoxaline
11095365
1,3-Dihydrothieno[3,4-b]chinoxalin
C1C2=NC3=CC=CC=C3N=C2CS1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3138-80-5
C10H8N2S
188.248
MFCD22683025
OSKBIJDQSJQRQK-UHFFFAOYSA-N
1,3-dihydrothieno 3,4-b quinoxaline, thieno 3,4-b quinoxaline, 1,3-dihydro, 1h,3h-thieno 3,4-b quinoxaline
11095365
1,3-Dihydrothieno[3,4-b]chinoxalin
C1C2=NC3=CC=CC=C3N=C2CS1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1,3-Dihydrothieno[3,4-b]quinoxaline
Menge 10 g
Formelmasse 188.25
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'