missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methoxybenzoyl)-2-(1-naphthoyl)hydrazin, 98 %

Catalog Number 11325219
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
73941-12-5
C19H16N2O3
320.348
MFCD00021528
MMJQUHDGIXJLMO-UHFFFAOYSA-N
m-anisic acid, 1-naphthoyl hydrazide, 1-m-methoxybenzoyl-2-1-naphthoyl hydrazine, hydrazine, 1-m-methoxybenzoyl-2-1-naphthoyl, 1-3-methoxybenzoyl-2-1-naphthoyl hydrazine, 2'-3-methoxybenzoyl-1-naphthohydrazide, 1-3-methoxybenzoyl-2-1-naphthoyl-hydrazine, 1-naphthalenecarboxylic acid, 2-3-methoxybenzoyl hydrazide, n'-3-methoxybenzoyl naphthalene-1-carbohydrazide, wln: l66j bvmmvr co1, n'-3-methoxybenzoyl-1-naphthohydrazide
52790
N'-(3-methoxybenzoyl)naphthalin-1-carbohydrazid
COC1=CC=CC(=C1)C(=O)NNC(=O)C2=CC=CC3=CC=CC=C32
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
73941-12-5
C19H16N2O3
320.348
MFCD00021528
MMJQUHDGIXJLMO-UHFFFAOYSA-N
m-anisic acid, 1-naphthoyl hydrazide, 1-m-methoxybenzoyl-2-1-naphthoyl hydrazine, hydrazine, 1-m-methoxybenzoyl-2-1-naphthoyl, 1-3-methoxybenzoyl-2-1-naphthoyl hydrazine, 2'-3-methoxybenzoyl-1-naphthohydrazide, 1-3-methoxybenzoyl-2-1-naphthoyl-hydrazine, 1-naphthalenecarboxylic acid, 2-3-methoxybenzoyl hydrazide, n'-3-methoxybenzoyl naphthalene-1-carbohydrazide, wln: l66j bvmmvr co1, n'-3-methoxybenzoyl-1-naphthohydrazide
52790
N'-(3-methoxybenzoyl)naphthalin-1-carbohydrazid
COC1=CC=CC(=C1)C(=O)NNC(=O)C2=CC=CC3=CC=CC=C32

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-(3-Methoxybenzoyl)-2-(1-naphthoyl)hydrazine
Schmelzpunkt 189°C to 191°C
Menge 1 g
Formelmasse 320.33
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'