missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetrakis(diethylaminocarbonylmethyl)-1,4,8,11-Tetraazacyclotetradecan, Thermo Scientific™

Catalog Number p-7028949
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
126320-56-7
C34H68N8O4
652.97
MFCD09263304
JKQNZLKTODZGSY-UHFFFAOYSA-N
tetamet2, 1,4,8,11-tetrakis diethylaminocarbonylmethyl-1,4,8,11-tetraazacyclotetradecane, acmc-20ap3f, cyclam-n,n',n',n-tetrakis n,n-diethylacetamide, 1,4,8,11-tetrakis diethylaminocarbonylmethyl-1,4,8,11-tetra, 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrakis n,n-diethylacetamide, 2,2',2,2'-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl tetrakis n,n-diethylacetamide, n,n-diethyl-2-4,8,11-tris diethylcarbamoyl methyl-1,4,8,11-tetraazacyclotetradecan-1-yl acetamide
14434737
N,N-Diethyl-2-[4,8,11-tris[2-(diethylamino)-2-oxoethyl]-1,4,8,11-tetrazacyclotetradec-1-yl]acetamid
CCN(CC)C(=O)CN1CCCN(CCN(CCCN(CC1)CC(=O)N(CC)CC)CC(=O)N(CC)CC)CC(=O)N(CC)CC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
126320-56-7
C34H68N8O4
652.97
MFCD09263304
JKQNZLKTODZGSY-UHFFFAOYSA-N
tetamet2, 1,4,8,11-tetrakis diethylaminocarbonylmethyl-1,4,8,11-tetraazacyclotetradecane, acmc-20ap3f, cyclam-n,n',n',n-tetrakis n,n-diethylacetamide, 1,4,8,11-tetrakis diethylaminocarbonylmethyl-1,4,8,11-tetra, 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrakis n,n-diethylacetamide, 2,2',2,2'-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl tetrakis n,n-diethylacetamide, n,n-diethyl-2-4,8,11-tris diethylcarbamoyl methyl-1,4,8,11-tetraazacyclotetradecan-1-yl acetamide
14434737
N,N-Diethyl-2-[4,8,11-tris[2-(diethylamino)-2-oxoethyl]-1,4,8,11-tetrazacyclotetradec-1-yl]acetamid
CCN(CC)C(=O)CN1CCCN(CCN(CCCN(CC1)CC(=O)N(CC)CC)CC(=O)N(CC)CC)CC(=O)N(CC)CC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1,4,8,11-Tetrakis(diethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
Schmelzpunkt 144°C to 145°C
Menge 250 mg
Empfindlichkeit Hygroscopic
Formelmasse 652.97
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'