Learn More
1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione, TRC
Product Code. 30473185
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473185 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30473185 Supplier Toronto Research Chemicals Supplier No. TRCM3206355G
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione
CAS 720-94-5
Molecular Formula C11 H9 F3 O2
Quantity 5 g
Synonym 1,3-Butanedione','4,4,4-trifluoro-1-(4-methylphenyl)-, 1,3-Butanedione','4,4,4-trifluoro-1-p-tolyl- (6CI,8CI), 4,4,4-Trifluoro-1-(4-methylphenyl)-1,3-butanedione, (4-Methylbenzoyl)trifluoroacetone, 1,1,1-Trifluoro-4-(4-methylphenyl)-2,4-butanedione, 1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione, 4-(4-Methylphenyl)-1,1,1-trifluorobutane-2,4-dione, 4-Methyl-1-(4,4,4-trifluoro-3-oxobutanoyl)benzene
InChI Formula InChI=1S/C11H9F3O2/c1-7-2-4-8(5-3-7)9(15)6-10(16)11(12,13)14/h2-5H,6H2,1H3
SMILES Cc1ccc(cc1)C(=O)CC(=O)C(F)(F)F
IUPAC Name 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione
Molecular Weight (g/mol) 230.18
Formula Weight 230.0555