missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylisatin, 97 %, Thermo Scientific Chemicals

Catalog Number p-7047097
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
574-17-4
C10H7NO3
189.17
MFCD00158542
LPGDEHBASRKTDG-UHFFFAOYSA-N
1-acetylisatin, n-acetylisatin, 1-acetyl-1h-indole-2,3-dione, acetylisatin, 1-acetylindoline-2,3-dione, 1-acetyl-indole-2,3-dione, 1h-indole-2,3-dione, 1-acetyl, indole-2,3-dione, 1-acetyl, 1-acetyl-2,3-dihydro-1h-indole-2,3-dione, n-acetyl-isatin
11321
CHEBI:16050
1-acetyl-2,3-dihydro-1H-indole-2,3-dione
CC(=O)N1C(=O)C(=O)C2=CC=CC=C12
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
574-17-4
C10H7NO3
189.17
MFCD00158542
LPGDEHBASRKTDG-UHFFFAOYSA-N
1-acetylisatin, n-acetylisatin, 1-acetyl-1h-indole-2,3-dione, acetylisatin, 1-acetylindoline-2,3-dione, 1-acetyl-indole-2,3-dione, 1h-indole-2,3-dione, 1-acetyl, indole-2,3-dione, 1-acetyl, 1-acetyl-2,3-dihydro-1h-indole-2,3-dione, n-acetyl-isatin
11321
CHEBI:16050
1-acetyl-2,3-dihydro-1H-indole-2,3-dione
CC(=O)N1C(=O)C(=O)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Acetylisatin
Schmelzpunkt 141°C to 143°C
Menge 1 g
Beilstein 113959
Formelmasse 189.17
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'