missing translation for 'onlineSavingsMsg'
Learn More

1-Benzensulfonyl-4-Brom-1H-Pyrazol, 97 %, Thermo Scientific™

Catalog Number p-7039873
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
121358-73-4
C9H7BrN2O2S
287.13
MFCD06080376
SAQVJLOSQUFTDU-UHFFFAOYSA-N
4-bromo-1-phenylsulfonyl pyrazole, 4-bromo-1-phenylsulfonyl-1h-pyrazole, 1-benzenesulfonyl-4-bromopyrazole, 1-benzenesulfonyl-4-bromo-1h-pyrazole, acmc-209aa2
6468394
1-(benzolsulfonyl)-4-brompyrazol
BrC1=CN(N=C1)S(=O)(=O)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
121358-73-4
C9H7BrN2O2S
287.13
MFCD06080376
SAQVJLOSQUFTDU-UHFFFAOYSA-N
4-bromo-1-phenylsulfonyl pyrazole, 4-bromo-1-phenylsulfonyl-1h-pyrazole, 1-benzenesulfonyl-4-bromopyrazole, 1-benzenesulfonyl-4-bromo-1h-pyrazole, acmc-209aa2
6468394
1-(benzolsulfonyl)-4-brompyrazol
BrC1=CN(N=C1)S(=O)(=O)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Benzenesulfonyl-4-bromo-1H-pyrazole
Menge 1 g
Formelmasse 287.13
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'