missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-Oxopiperazin, 95 %, Thermo Scientific™

Catalog Number p-7036731
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
13754-41-1
C11H14N2O
190.246
MFCD06795962
SBWVHKNCFZRBRJ-UHFFFAOYSA-N
1-benzyl-3-oxopiperazine, 4-benzyl-piperazin-2-one, piperazinone, 4-phenylmethyl, 2-piperazinone,4-phenylmethyl, 4-benzyl-2-piperazinone, 4-benzylpiperazine-2-one, 4-phenylmethyl piperazinone, aronis25032
11458177
4-Benzylpiperazin-2-on
C1CN(CC(=O)N1)CC2=CC=CC=C2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
13754-41-1
C11H14N2O
190.246
MFCD06795962
SBWVHKNCFZRBRJ-UHFFFAOYSA-N
1-benzyl-3-oxopiperazine, 4-benzyl-piperazin-2-one, piperazinone, 4-phenylmethyl, 2-piperazinone,4-phenylmethyl, 4-benzyl-2-piperazinone, 4-benzylpiperazine-2-one, 4-phenylmethyl piperazinone, aronis25032
11458177
4-Benzylpiperazin-2-on
C1CN(CC(=O)N1)CC2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Benzyl-3-oxopiperazine
Schmelzpunkt 148°C to 155°C
Menge 250 mg
Formelmasse 190.24
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'