missing translation for 'onlineSavingsMsg'
Learn More

1-(Benzyloxycarbonylamino)cyclobutancarbonsäure, 97%, Thermo Scientific™

Catalog Number p-7041444
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
190004-53-6
C13H15NO4
249.27
MFCD03426163
GDJSFBNRXFOUEQ-UHFFFAOYSA-N
1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, 1-z-amino cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid, z-ac4c-oh, 1-benzyloxycarbonylamino-cyclobutyl-1-carboxylic acid, 1-benzyloxy carbonyl amino-cyclobutanecarboxylic acid, z-cyclovaline, z-acbc-oh, 1-benzyloxycarbonylaminocyclobutanecarboxylic acid
1512648
1-(Phenylmethoxycarbonylamino)cyclobutan-1-carbonsäure
OC(=O)C1(CCC1)NC(=O)OCC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
190004-53-6
C13H15NO4
249.27
MFCD03426163
GDJSFBNRXFOUEQ-UHFFFAOYSA-N
1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, 1-z-amino cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid, z-ac4c-oh, 1-benzyloxycarbonylamino-cyclobutyl-1-carboxylic acid, 1-benzyloxy carbonyl amino-cyclobutanecarboxylic acid, z-cyclovaline, z-acbc-oh, 1-benzyloxycarbonylaminocyclobutanecarboxylic acid
1512648
1-(Phenylmethoxycarbonylamino)cyclobutan-1-carbonsäure
OC(=O)C1(CCC1)NC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
Schmelzpunkt 83°C to 87°C
Formelmasse 249.26
Reinheit (%) 97%
Menge 250 mg
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'