Learn More
1-Brom-3-(difluormethoxy)-Benzol, 97+ %, Thermo Scientific™
Catalog Number p-7027314
Click to view available options
Menge:
1 g
5 g
262587-05-3
C7H5BrF2O
223.017
MFCD00236233
NWBDGKNKAFGQQL-UHFFFAOYSA-N
1-bromo-3-difluoromethoxy benzene, 3-difluoromethoxy bromobenzene, benzene, 1-bromo-3-difluoromethoxy, 1-difluoromethoxy-3-bromobenzene, pubchem18537, acmc-1crzp, 3-difluoromethoxybromobenzene, ksc493c1b, 3-bromo-1-difluoromethoxy benzene, 3-difluoromethyl bromobenzene
2737006
1-Brom-3-(difluormethoxy)benzol
C1=CC(=CC(=C1)Br)OC(F)F
262587-05-3
C7H5BrF2O
223.017
MFCD00236233
NWBDGKNKAFGQQL-UHFFFAOYSA-N
1-bromo-3-difluoromethoxy benzene, 3-difluoromethoxy bromobenzene, benzene, 1-bromo-3-difluoromethoxy, 1-difluoromethoxy-3-bromobenzene, pubchem18537, acmc-1crzp, 3-difluoromethoxybromobenzene, ksc493c1b, 3-bromo-1-difluoromethoxy benzene, 3-difluoromethyl bromobenzene
2737006
1-Brom-3-(difluormethoxy)benzol
C1=CC(=CC(=C1)Br)OC(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Bromo-3-(difluoromethoxy)benzene
Menge 1 g
Formelmasse 223.02
Reinheit (%) ≥97%