missing translation for 'onlineSavingsMsg'
Learn More

1-Brom-4-Chlor-2-Nitrobenzol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7020617
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
41513-04-6
C6H3BrClNO2
236.449
MFCD00024320
UKTIMFAJRPSNGR-UHFFFAOYSA-N
2-bromo-5-chloronitrobenzene, benzene, 1-bromo-4-chloro-2-nitro, 4-bromo-3-nitrochlorobenzene, 3-chloro-6-bromonitrobenzene, pubchem3593, acmc-209jkn, 1-bromo-4-chloronitrobenzene, ksc490m7r, 2-bromo-5-chloro-nitrobenzene, bromo-4-chloro-2-nitrobenzene
2794904
1-Brom-4-Chlor-2-nitrobenzol
C1=CC(=C(C=C1Cl)[N+](=O)[O-])Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
41513-04-6
C6H3BrClNO2
236.449
MFCD00024320
UKTIMFAJRPSNGR-UHFFFAOYSA-N
2-bromo-5-chloronitrobenzene, benzene, 1-bromo-4-chloro-2-nitro, 4-bromo-3-nitrochlorobenzene, 3-chloro-6-bromonitrobenzene, pubchem3593, acmc-209jkn, 1-bromo-4-chloronitrobenzene, ksc490m7r, 2-bromo-5-chloro-nitrobenzene, bromo-4-chloro-2-nitrobenzene
2794904
1-Brom-4-Chlor-2-nitrobenzol
C1=CC(=C(C=C1Cl)[N+](=O)[O-])Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Bromo-4-chloro-2-nitrobenzene
Schmelzpunkt 68°C to 72°C
Menge 5 g
Beilstein 1950424
Formelmasse 236.45
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'