missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluorobenzene, 99%

Product Code. 11414733
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414733 2500 g 2500g
11494723 100 g 100g
11404733 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414733 Supplier Thermo Scientific Alfa Aesar Supplier No. A11630.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 460-00-4 | C6H4BrF | 175.00 g/mol

1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

Solubility
Soluble in water(0.14g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 460-00-4
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175.00
MDL Number MFCD00000342
InChI Key AITNMTXHTIIIBB-UHFFFAOYSA-N
Synonym 4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
PubChem CID 9993
IUPAC Name 1-bromo-4-fluorobenzene
SMILES FC1=CC=C(Br)C=C1

Specifications

Melting Point -16°C
Density 1.604
Boiling Point 151°C to 153°C
Flash Point 60°C (140°F)
Odor Aromatic
Refractive Index 1.527
Quantity 2500 g
UN Number UN1993
Beilstein 774102
Solubility Information Soluble in water(0.14g/L).
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.