Learn More
1-Chloroethyl Isopropyl Carbonate, TRC
Product Code. 30367142
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367142 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30367142 Supplier Toronto Research Chemicals Supplier No. TRCC984675500MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Chloroethyl Isopropyl Carbonate
CAS 98298-66-9
Molecular Formula C6H11O3Cl
Quantity 500 mg
InChI Formula InChI=1S/C6H11ClO3/c1-4(2)9-6(8)10-5(3)7/h4-5H,1-3H3
SMILES CC(C)OC(=O)OC(C)Cl
IUPAC Name 1-chloroethyl propan-2-yl carbonate
Molecular Weight (g/mol) 166.6
Formula Weight 166.0397