missing translation for 'onlineSavingsMsg'
Learn More

1-Decylboronsäure, 98 %, Thermo Scientific Chemicals

Catalog Number p-7047916
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24464-63-9
C10H23BO2
186.102
MFCD01074638
TUTSEMMBKGBUGD-UHFFFAOYSA-N
n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d
4186635
Decylboronsäure
B(CCCCCCCCCC)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24464-63-9
C10H23BO2
186.102
MFCD01074638
TUTSEMMBKGBUGD-UHFFFAOYSA-N
n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d
4186635
Decylboronsäure
B(CCCCCCCCCC)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Decylboronic acid
Schmelzpunkt 86°C to 89°C
Menge 1 g
Beilstein 1743934
Formelmasse 186.1
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'