missing translation for 'onlineSavingsMsg'
Learn More

1-Deoxy-1-nitro-L-iditolhemihydrat, 99 %, Thermo Scientific™

Catalog Number p-7025889
Click to view available options
Menge:
100 mg
500 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
207226-23-1
C12H28N2O15
440.36
MFCD00191972
HQAUHMYCAWVBKQ-UHFFFAOYNA-N
1-deoxy-1-nitro-l-iditol hemihydrate, 2s,3r,4r,5s-6-nitrohexane-1,2,3,4,5-pentol hydrate
73995060
(2S,3R,4R,5S)-6-Nitrohexan-1,2,3,4,5-Pentol;hydrat
O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
207226-23-1
C12H28N2O15
440.36
MFCD00191972
HQAUHMYCAWVBKQ-UHFFFAOYNA-N
1-deoxy-1-nitro-l-iditol hemihydrate, 2s,3r,4r,5s-6-nitrohexane-1,2,3,4,5-pentol hydrate
73995060
(2S,3R,4R,5S)-6-Nitrohexan-1,2,3,4,5-Pentol;hydrat
O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Deoxy-1-nitro-L-iditol hemihydrate
Schmelzpunkt 87°C to 89°C
Menge 100 mg
Formelmasse 220.19 (211.18 Anhydrous)
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'