missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium Hexafluorphosphat, 98 %, Thermo Scientific™

Catalog Number p-7040452
Click to view available options
Menge:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
292140-86-4
C7H13F6N2P
270.159
MFCD08458503
XLKYEJKYOZNSDQ-UHFFFAOYSA-N
1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, dsstox_cid_29120, dsstox_rid_83339, dsstox_gsid_49264, 1-ethyl-2,3-dimethylimidazol-1-ium hexafluorophosphate
50878556
1-Ethyl-2,3-Dimethylimidazol-3-ium;Hexafluorphosphat
CCN1C=C[N+](=C1C)C.F[P-](F)(F)(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
292140-86-4
C7H13F6N2P
270.159
MFCD08458503
XLKYEJKYOZNSDQ-UHFFFAOYSA-N
1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, dsstox_cid_29120, dsstox_rid_83339, dsstox_gsid_49264, 1-ethyl-2,3-dimethylimidazol-1-ium hexafluorophosphate
50878556
1-Ethyl-2,3-Dimethylimidazol-3-ium;Hexafluorphosphat
CCN1C=C[N+](=C1C)C.F[P-](F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-Ethyl-2,3-dimethylimidazolium hexafluorophosphate
Menge 10 g
Formelmasse 270.16
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'